Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-p-anisaldehyde ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

009760-25G 009760-100G 009760-500G
101797-182EA 61.54 USD InStock
101797-182 101797-184 101797-186
3-Bromo-p-anisaldehyde ≥98% for synthesis
3-Bromo-p-anisaldehyde
Hazard code(s): Irritant, Air Sensitive


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 108…110 °C (1 mmHg)
Melting Pt: 54
MDL Number: MFCD00016599
CAS Number: 34841-06-0
EINECS: 252-241-8
Order Now
Specification Test Results
Melting point 51-53°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty