Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-pyridineboronic acid hydrate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

009785-5G 009785-25G
101797-250EA 21.54 USD InStock
101797-250 101797-252
2-Fluoro-3-pyridineboronic acid hydrate ≥98% for synthesis
2-Fluoro-3-pyridineboronic acid hydrate


Formula: C₅H₅BFNO₂·xH₂O
MW: 140.91 g/mol (anhy)
MDL Number: MFCD04038758
CAS Number: 174669-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty