Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

011109-25G 011109-100G
101799-876EA 38.46 USD InStock
101799-876 101799-878
2,4-Dichloropyridine ≥98%
2,4-Dichloropyridine
Hazard code(s): Irritant


Formula: C₅H₃Cl₂N
Boiling Pt: 189…190 °C
Melting Pt: –1–0 °C
MDL Number: MFCD00955618
CAS Number: 26452-80-2
EINECS: 247-717-7
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Boiling point 189-190°C
Melting point -1-0°C
ND20 1.5485

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty