Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

011115-25G 011115-100G
101799-898EA 49.23 USD InStock
101799-898 101799-900
3-Bromo-2-chloropyridine ≥98%
3-Bromo-2-chloropyridine
Hazard code(s): Irritant


Formula: C₅H₃BrClN
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 54…57 °C
MDL Number: MFCD00234007
CAS Number: 52200-48-3
Order Now
Specification Test Results
Melting point 54-57°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty