Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-bromopyridine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

011118-5G 011118-25G
101799-912EA 36.92 USD InStock
101799-912 101799-914
3-Amino-2-bromopyridine ≥98% for synthesis
3-Amino-2-bromopyridine
Hazard code(s): Irritant


Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 78…81 °C
MDL Number: MFCD00234064
CAS Number: 39856-58-1
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 78-81°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty