Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dichloroisonicotinic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

011178-5G 011178-25G
101800-090EA 33.85 USD InStock
101800-090 101800-092
2,6-Dichloroisonicotinic acid ≥98% for synthesis
2,6-Dichloroisonicotinic acid
Hazard code(s): Irritant


Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 208 °C (dec.)
MDL Number: MFCD00234147
CAS Number: 5398-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty