Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluoropyridine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Danger

011261-1G 011261-5G 011261-25G
101800-260EA 26.15 USD InStock
101800-260 101800-262 101800-264
4-Chloro-3-fluoropyridine ≥98% for synthesis
4-Chloro-3-fluoropyridine
Hazard code(s): Irritant


Formula: C₅H₃ClFN
MW: 131.54
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty