Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyindole ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

011268-1G 011268-5G
101800-278EA 29.23 USD InStock
101800-278 101800-280
6-Methoxyindole ≥98% for synthesis
6-Methoxyindole
Hazard code(s): Irritant, Light Sensitive


Formula: C₉H₉NO
Boiling Pt: 105 °C (0.2mm)
Melting Pt: 90…94 °C
Storage Temperature: Freezer
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6
Order Now
Specification Test Results
Melting point 90-94°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty