Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-α,α,α,4-tetrafluorotoluene ≥98%

Supplier: MATRIX SCIENTIFIC MS

011286-5G 011286-25G
101800-338EA 50.77 USD InStock
101800-338 101800-340
2-Bromo-α,α,α,4-tetrafluorotoluene ≥98%
2-Bromo-α,α,α,4-tetrafluorotoluene
Hazard code(s): Irritant


Formula: C₇H₃BrF₄
Boiling Pt: 161…162 °C
Density: 1.695 g/cm³
MDL Number: MFCD03094041
CAS Number: 351003-21-9
Order Now
Specification Test Results
Boiling point 161-162°C
Density 1.695
ND20 1.465

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty