Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromopicolinic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

012526-25G 012526-100G
101802-764EA 84.62 USD InStock
101802-764 101802-766
5-Bromopicolinic acid ≥98% for synthesis
5-Bromopicolinic acid
Hazard code(s): Irritant


Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 173…175 °C
MDL Number: MFCD00234149
CAS Number: 30766-11-1
Order Now
Specification Test Results
Melting point 173-175°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty