Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-4-carbaldehyd ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

012528-5G 012528-25G
101802-772EA 89.23 USD InStock
101802-772 101802-774
Indole-4-carbaldehyd ≥98% for synthesis
Indole-4-carbaldehyd
Hazard code(s): Irritant, Air Sensitive, Store in Freezer


Formula: C₉H₇NO
MW: 145.16
Melting Pt: 143…144 °C
MDL Number: MFCD01632221
CAS Number: 1074-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty