Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-2-thenoic acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

012535-25G 012535-100G
101802-790EA 44.62 USD InStock
101802-790 101802-792
3-Methyl-2-thenoic acid ≥98%
3-Methyl-2-thenoic acid
Hazard code(s): Irritant


Formula: C₆H₆O₂S
Melting Pt: 143…144 °C
MDL Number: MFCD00005438
CAS Number: 23806-24-8
EINECS: 245-894-5
Order Now
Specification Test Results
Melting point 143-144°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty