Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-4-Bromomandelic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

012636-5G 012636-25G
101803-048EA 18.46 USD InStock
101803-048 101803-050
(±)-4-Bromomandelic acid ≥98% for synthesis
(±)-4-Bromomandelic acid
Hazard code(s): Irritant


Formula: C₈H₇BrO₃
MW: 231.05
Boiling Pt: 344
Melting Pt: 116…118 °C
MDL Number: MFCD00004232
CAS Number: 6940-50-7
EINECS: 230-085-1
Merck Index: 13,01409
Order Now
Specification Test Results
Melting point 116-118°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty