Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl indole-6-carboxylate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

012664-5G 012664-25G
101803-144EA 27.69 USD InStock
101803-144 101803-146
Methyl indole-6-carboxylate ≥98% for synthesis
Methyl indole-6-carboxylate
Hazard code(s): Irritant, Store in Freezer


Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00211063
CAS Number: 50820-65-0
Order Now
Specification Test Results
Melting point 78-80°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty