Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-iodobenzoate ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

012668-25G 012668-100G
101803-158EA 44.62 USD InStock
101803-158 101803-160
Methyl-3-iodobenzoate ≥98% for synthesis
Methyl-3-iodobenzoate
Hazard code(s): Irritant


Formula: C₈H₇IO₂
MW: 262.05
Melting Pt: 46…50 °C
MDL Number: MFCD00061093
CAS Number: 618-91-7
EINECS: 210-570-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty