Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromopyridine-2-carbonitrile for synthesis

Supplier: MATRIX SCIENTIFIC MS

018246-1G 018246-25G
101805-720EA 61.54 USD InStock
101805-720 102663-606
3-Bromopyridine-2-carbonitrile for synthesis
3-Bromopyridine-2-carbonitrile
Hazard code(s): Irritant


Formula: C₆H₃BrN₂
MW: 183.01
Boiling Pt: 120 °C (4mm)
Melting Pt: 98.2…99.0 °C
MDL Number: MFCD02683288
CAS Number: 55758-02-6
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 120°C/4mm
Melting point 98.2-99.0°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty