Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-bromopyridine ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

020568-25G 020568-100G
101810-308EA 90.77 USD InStock
101810-308 102663-622
5-Amino-2-bromopyridine ≥97% for synthesis
5-Amino-2-bromopyridine
Hazard code(s): Irritant


Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 76…77 °C
MDL Number: MFCD01646116
CAS Number: 13534-97-9
Order Now
Specification Test Results
Melting point 76-77°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty