Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromo-3-picoline ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

021366-5G 021366-25G 021366-100G
101811-860EA 16.92 USD InStock
101811-860 101811-862 101811-864
2,5-Dibromo-3-picoline ≥98% for synthesis
2,5-Dibromo-3-picoline
Hazard code(s): Irritant


Formula: C₆H₅Br₂N
MW: 250.92
Melting Pt: 43…47 °C
MDL Number: MFCD02093085
CAS Number: 3430-18-0
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 43-47°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty