Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoroanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

021386-5G 021386-25G
101811-928EA 15.38 USD InStock
101811-928 101811-930
5-Bromo-2-fluoroanisole ≥98%
5-Bromo-2-fluoroanisole
Hazard code(s): Irritant


Formula: C₇H₆BrFO
MDL Number: MFCD01861130
CAS Number: 103291-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty