Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-5-nitropyrimidine ≥97%

Supplier: MATRIX SCIENTIFIC MS
Warning

021422-25G 021422-100G
101812-056EA 72.31 USD InStock
101812-056 101812-058
2,4-Dichloro-5-nitropyrimidine ≥97%
2,4-Dichloro-5-nitropyrimidine
Hazard code(s): Irritant, Lachrymator, Avoid Skin Contact, Store in Freezer


Formula: C₄HCl₂N₃O₂
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00127867
CAS Number: 49845-33-2
Order Now
Specification Test Results
Melting point 28-32°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty