Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)benzenesulphonamide ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

021428-5G 021428-25G
101812-074EA 18.46 USD InStock
101812-074 101812-076
4-(Trifluoromethyl)benzenesulphonamide ≥97% for synthesis
4-(Trifluoromethyl)benzenesulphonamide
Hazard code(s): Irritant


Formula: C₇H₆F₃NO₂S
MW: 225.19
Melting Pt: 175…180 °C
MDL Number: MFCD00159251
CAS Number: 830-43-3
EINECS: 212-596-1
Order Now
Specification Test Results
Melting point 175-180°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty