Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chloro-3-methylpyridine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

023759-5G 023759-25G
101816-660EA 21.54 USD InStock
101816-660 101816-662
2-Amino-5-chloro-3-methylpyridine ≥98% for synthesis
2-Amino-5-chloro-3-methylpyridine
Hazard code(s): Irritant


Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD00129030
CAS Number: 20712-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty