Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluorobenzonitrile ≥98%

Supplier: MATRIX SCIENTIFIC MS
Warning

023805-5G 023805-25G
101816-794EA 23.08 USD InStock
101816-794 101816-796
2-Bromo-5-fluorobenzonitrile ≥98%
2-Bromo-5-fluorobenzonitrile
Hazard code(s): Irritant-Harmful


Formula: C₇H₃BrFN
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89 - 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00142875
CAS Number: 57381-39-2
EINECS: 260-711-9
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 89-91°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty