Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-p-toluic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

024051-25G 024051-100G
101817-306EA 46.15 USD InStock
101817-306 101817-308
3-Bromo-p-toluic acid ≥97% for synthesis
3-Bromo-p-toluic acid
Hazard code(s): Irritant


Formula: C₈H₇BrO₂
MW: 215.05
Melting Pt: 206…208 °C
MDL Number: MFCD00002488
CAS Number: 7697-26-9
EINECS: 231-712-1
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 206-208°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty