Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-nitro-4-picoline ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

024590-5G 024590-25G
101818-382EA 49.23 USD InStock
101818-382 101818-384
2-Amino-5-nitro-4-picoline ≥98% for synthesis
2-Amino-5-nitro-4-picoline
Hazard code(s): Irritant


Formula: C₆H₇N₃O₂
MW: 153.14
Melting Pt: 220
MDL Number: MFCD00010692
CAS Number: 21901-40-6
Order Now
Specification Test Results
Melting point 223-225°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty