Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-difluorobenzoic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

024609-25G 024609-100G
101818-424EA 81.54 USD InStock
101818-424 101818-426
4-Bromo-2,6-difluorobenzoic acid ≥98% for synthesis
4-Bromo-2,6-difluorobenzoic acid
Hazard code(s): Irritant


Formula: C₇H₃BrF₂O₂
MW: 237
Melting Pt: 201…203 °C
MDL Number: MFCD03094085
CAS Number: 183065-68-1
EINECS: 457-940-3
Order Now
Specification Test Results
Melting point 201-203°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty