Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-5-nitropyridine ≥98%

Supplier: MATRIX SCIENTIFIC MS

024645-5G 024645-25G
101818-544EA 36.92 USD InStock
101818-544 101818-546
3-Bromo-2-chloro-5-nitropyridine ≥98%
3-Bromo-2-chloro-5-nitropyridine
Hazard code(s): Irritant


Formula: C₅H₂BrClN₂O₂
Melting Pt: 67…68 °C
MDL Number: MFCD00233989
CAS Number: 5470-17-7
Order Now
Specification Test Results
Melting point 67-68°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty