Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromophenylboronic acid ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

024996-5G 024996-25G 024996-100G
101819-366EA 58.46 USD InStock
101819-366 101819-368 102663-650
2-Bromophenylboronic acid ≥98% for synthesis
2-Bromophenylboronic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD01114672
CAS Number: 244205-40-1
Order Now
Specification Test Results
Melting point 108-110°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty