Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(trifluoromethyl)pyrimidine ≥98% for synthesis

Supplier: MATRIX SCIENTIFIC MS

026309-1G 026309-5G
101822-056EA 47.69 USD InStock
101822-056 101822-058
2-Amino-4-(trifluoromethyl)pyrimidine ≥98% for synthesis
2-Amino-4-(trifluoromethyl)pyrimidine
Hazard code(s): Irritant


Formula: C₅H₄F₃N₃
MW: 163.1
Melting Pt: 175…177 °C
MDL Number: MFCD00828624
CAS Number: 16075-42-6
Order Now
Specification Test Results
Melting point 175-177°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty