Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butylpiperazine ≥95%

Supplier: MATRIX SCIENTIFIC MS

029394-5G 029394-25G
101828-066EA 138.46 USD InStock
101828-066 101828-068
1-Butylpiperazine ≥95%
1-Butylpiperazine
Hazard code(s): Irritant


Formula: C₈H₁₈N₂
MDL Number: MFCD00039702
CAS Number: 5610-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty