Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-o-toluic acid ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

032351-25G 032351-100G
101833-536EA 72.31 USD InStock
101833-536 101833-538
4-Bromo-o-toluic acid ≥97% for synthesis
4-Bromo-o-toluic acid
Hazard code(s): Toxic


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 186…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2
EINECS: 272-437-7
Order Now
Specification Test Results
Melting point 186-187°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty