Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethoxyaniline ≥95%

Supplier: MATRIX SCIENTIFIC MS

034260-1G 034260-5G 034260-25G
101837-296EA 40 USD InStock
101837-296 101837-298 102663-670
2,6-Dimethoxyaniline ≥95%
2,6-Dimethoxyaniline
Hazard code(s): Toxic, Irritant


Formula: C₈H₁₁NO₂
MDL Number: MFCD00053934
CAS Number: 2734-70-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty