Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

035391-1G 035391-5G
101839-394EA 15.38 USD InStock
101839-394 101839-396
2-Chloro-5-(trifluoromethyl)phenylboronic acid ≥95% for synthesis
2-Chloro-5-(trifluoromethyl)phenylboronic acid
Hazard code(s): Irritant, Store in Freezer


Formula: C₇H₅BClF₃O₂
MW: 224.37
MDL Number: MFCD00797335
CAS Number: 182344-18-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty