Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxytryptamine hydrochloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

036774-5G 036774-25G
101842-054EA 27.69 USD InStock
101842-054 101842-056
5-Methoxytryptamine hydrochloride ≥98%
5-Methoxytryptamine hydrochloride
Hazard code(s): Irritant


Formula: C₁₁H₁₅ClN₂O
Melting Pt: 121…123 °C
MDL Number: MFCD00005662
CAS Number: 66-83-1
EINECS: 200-637-6
Order Now
Specification Test Results
Melting point 121-123 ºC

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty