Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isopropylbenzylamine ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

037451-100G 037451-500G
101843-408EA 120 USD InStock
101843-408 101843-410
N-Isopropylbenzylamine ≥95% for synthesis
N-Isopropylbenzylamine
Hazard code(s): Irritant


Formula: C₆H₅CH₂N(CH₃)(CH₂CH₃)
MW: 149.24 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: ∼ 0 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008863
CAS Number: 102-97-6
EINECS: 203-067-6
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty