Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyphenyl isocyanate

Supplier: MATRIX SCIENTIFIC MS

038320-1G 038320-5G
101845-266EA 16.92 USD InStock
101845-266 101845-268
2-Methoxyphenyl isocyanate
2-Methoxyphenyl isocyanate
Hazard code(s): Toxic, Moisture Sensitive


Formula: C₈H₇NO₂
Boiling Pt: 94.7 °C (1013 hPa)
Density: 1.122 g/cm³ (20 °C)
MDL Number: MFCD00002004
CAS Number: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty