Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Tryptophan ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS
Warning

038632-25G 038632-100G
101845-896EA 56.92 USD InStock
101845-896 101845-898
DL-Tryptophan ≥97% for synthesis
DL-Tryptophan


Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 280…283 °C
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 205-819-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty