Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-o-anisic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

042648-500MG 042648-1G 042648-5G
101856-792EA 16.92 USD InStock
101856-792 101856-794 101856-796
4-Amino-o-anisic acid ≥95% for synthesis
4-Amino-o-anisic acid
Hazard code(s): Irritant


Formula: C₈H₉NO₃
Melting Pt: 136…137 °C
MDL Number: MFCD00114479
CAS Number: 2486-80-8
EINECS: 219-632-5
Order Now
Specification Test Results
Melting point 136-137°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty