Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromonicotinic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

043121-25G 043121-100G
101859-278EA 149.23 USD InStock
101859-278 102663-710
2-Amino-5-bromonicotinic acid ≥95% for synthesis
2-Amino-5-bromonicotinic acid
Hazard code(s): Irritant


Formula: C₆H₅BrN₂O₂
Boiling Pt: 350 °C
Melting Pt: 253…255 °C
Density: 1.909 g/cm³
MDL Number: MFCD01860227
CAS Number: 52833-94-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty