Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-chloro-5-(trifluoromethyl)picolinate ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

043387-1G 043387-5G
101860-534EA 46.15 USD InStock
101860-534 101860-536
Ethyl-3-chloro-5-(trifluoromethyl)picolinate ≥95% for synthesis
Ethyl 3-chloro-5-(trifluoromethyl)picolinate
Hazard code(s): Irritant


Formula: C₉H₇ClF₃NO₂
Boiling Pt: 176 °C (1013 hPa)
MDL Number: MFCD06656414
CAS Number: 128073-16-5
EINECS: 410-340-5
UN: 3082
ADR: 9,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty