Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Bromobiphenyl-4-carboxylic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

044247-500MG 044247-1G 044247-5G
101864-926EA 230.77 USD InStock
101864-926 101864-928 101864-930
4'-Bromobiphenyl-4-carboxylic acid ≥95% for synthesis
4'-Bromobiphenyl-4-carboxylic acid
Hazard code(s): Irritant


Formula: C₁₃H₉BrO₂
Melting Pt: 301…302 °C
MDL Number: MFCD00452920
CAS Number: 5731-11-3
Order Now
Specification Test Results
Melting point 301-302°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty