Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloroacetophenone oxime ≥95%

Supplier: MATRIX SCIENTIFIC MS

044394-500MG 044394-1G 044394-5G
101865-606EA 181.54 USD InStock
101865-606 101865-608 101865-610
4'-Chloroacetophenone oxime ≥95%
4'-Chloroacetophenone oxime
Hazard code(s): Irritant


Formula: C₈H₈ClNO
Melting Pt: 96…98 °C
MDL Number: MFCD02609517
CAS Number: 1956-39-4
Order Now
Specification Test Results
Melting point 96-98°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty