Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Toluoyl)benzoic acid ≥95%

Supplier: MATRIX SCIENTIFIC MS

044956-500MG 044956-1G 044956-5G
101868-426EA 181.54 USD InStock
101868-426 101868-428 101868-430
2-(4-Toluoyl)benzoic acid ≥95%
2-(4-Toluoyl)benzoic acid
Hazard code(s): Irritant


Formula: C₁₅H₁₂O₃
Melting Pt: 145…146 °C
MDL Number: MFCD00020287
CAS Number: 85-55-2
EINECS: 201-614-3
Order Now
Specification Test Results
Melting point 145-146°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty