Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3,5-diaminobenzoate ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

045039-500MG 045039-1G 045039-5G
101868-846EA 86.15 USD InStock
101868-846 101868-848 101868-850
Methyl-3,5-diaminobenzoate ≥95% for synthesis
Methyl-3,5-diaminobenzoate
Hazard code(s): Irritant


Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 133…135 °C
MDL Number: MFCD00041433
CAS Number: 1949-55-9
Order Now
Specification Test Results
Melting point 133-135°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty