Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloronicotinic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

045444-1G 045444-5G 045444-25G
101870-856EA 15.38 USD InStock
101870-856 101870-858 102833-136
2,5-Dichloronicotinic acid ≥95% for synthesis
2,5-Dichloronicotinic acid
Hazard code(s): Irritant


Formula: C₆H₃Cl₂NO₂
MW: 192
Melting Pt: 156…158 °C
MDL Number: MFCD01861979
CAS Number: 59782-85-3
Order Now
Specification Test Results
Melting point 156-158°C

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty