Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methylcaproic acid ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

045693-500MG 045693-1G 045693-5G
101872-138EA 104.62 USD InStock
101872-138 101872-140 101872-142
3-Amino-4-methylcaproic acid ≥95% for synthesis
3-Amino-4-methylcaproic acid
Hazard code(s): Irritant


Formula: C₇H₁₅NO₂
MW: 145.2
Melting Pt: 190…192 °C
MDL Number: MFCD03305808
CAS Number: 40469-87-2
Order Now
Specification Test Results
Melting point 190-192°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty