Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-methoxy-4-nitrobenzoate ≥95%

Supplier: MATRIX SCIENTIFIC MS

046159-500MG 046159-1G 046159-5G
101874-448EA 181.54 USD InStock
101874-448 101874-450 101874-452
Methyl-3-methoxy-4-nitrobenzoate ≥95%
Methyl-3-methoxy-4-nitrobenzoate
Hazard code(s): Irritant


Formula: C₉H₉NO₅
Melting Pt: 91…93 °C
MDL Number: MFCD00051967
CAS Number: 5081-37-8
Order Now
Specification Test Results
Melting point 91-93°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty