Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methylbenzothiazole ≥97% for synthesis

Supplier: MATRIX SCIENTIFIC MS

047084-500MG 047084-1G 047084-5G
101878-384EA 278.46 USD InStock
101878-384 101878-386 101878-388
2-Chloro-6-methylbenzothiazole ≥97% for synthesis
2-Chloro-6-methylbenzothiazole
Hazard code(s): Irritant


Formula: C₈H₆ClNS
MDL Number: MFCD04448821
CAS Number: 3507-26-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty