Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitroanisole ≥95%

Supplier: MATRIX SCIENTIFIC MS

047946-25G 047946-100G
101882-104EA 50.77 USD InStock
101882-104 101882-106
5-Fluoro-2-nitroanisole ≥95%
5-Fluoro-2-nitroanisole
Hazard code(s): Irritant


Formula: C₇H₆FNO₃
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00077541
CAS Number: 448-19-1
Order Now
Specification Test Results
Melting point 58-60°C

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty