Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-chloro-2-nitrobenzoate ≥95% for synthesis

Supplier: MATRIX SCIENTIFIC MS

047997-500MG 047997-1G 047997-5G
101882-324EA 181.54 USD InStock
101882-324 101882-326 101882-328
Methyl-5-chloro-2-nitrobenzoate ≥95% for synthesis
Methyl-5-chloro-2-nitrobenzoate


Formula: C₈H₆ClNO₄
Boiling Pt: 306 °C (1013 hPa)
Density: 1.426 g/cm³ (20 °C)
MDL Number: MFCD00272139
CAS Number: 51282-49-6
EINECS: 257-107-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty